Product Details

SNP ID
rs12482654
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:42500382 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGGATAGAGACAGACCCGAGATTG[C/G]AGCCCTAGCTCCGTATACTTCTTTC
Phenotype
MIM: 609314 MIM: 608094
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101930094 PubMed Links

Gene Details

Gene
LOC101930094
Gene Name
uncharacterized LOC101930094
There are no transcripts associated with this gene.

Gene
RSPH1
Gene Name
radial spoke head 1 homolog
There are no transcripts associated with this gene.

Gene
SLC37A1
Gene Name
solute carrier family 37 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320537.1 Intron NP_001307466.1
NM_018964.3 Intron NP_061837.3
XM_011529614.2 Intron XP_011527916.1
XM_011529615.1 Intron XP_011527917.1
XM_017028376.1 Intron XP_016883865.1
XM_017028377.1 Intron XP_016883866.1
XM_017028378.1 Intron XP_016883867.1
XM_017028379.1 Intron XP_016883868.1
XM_017028380.1 Intron XP_016883869.1
XM_017028381.1 Intron XP_016883870.1
XM_017028382.1 Intron XP_016883871.1
XM_017028383.1 Intron XP_016883872.1

View Full Product Details