Product Details

SNP ID
rs35462782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42845672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCTTCCAAGTCGATCATGAGC[C/T]GCCACAGCCGCAAACACTAGGCGTT
Phenotype
MIM: 605924
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR4 PubMed Links

Gene Details

Gene
WDR4
Gene Name
WD repeat domain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260474.1 Intron NP_001247403.1
NM_001260475.1 Intron NP_001247404.1
NM_001260476.1 Intron NP_001247405.1
NM_001260477.1 Intron NP_001247406.1
NM_018669.5 Intron NP_061139.2
NM_033661.4 Intron NP_387510.1
XM_011529433.1 Intron XP_011527735.1
XM_017028262.1 Intron XP_016883751.1
XM_017028263.1 Intron XP_016883752.1
XM_017028264.1 Intron XP_016883753.1

View Full Product Details