Product Details

SNP ID
rs354020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49335634 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTGAGTTGCCTCATGGCTACCCAG[A/C]CGGGGCCCAGCCCCTGCCGCTAACC
Phenotype
MIM: 151523 MIM: 607972 MIM: 601729
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD37 PubMed Links

Gene Details

Gene
CD37
Gene Name
CD37 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040031.1 Intron NP_001035120.1
NM_001774.2 Intron NP_001765.1
XM_005259435.3 Intron XP_005259492.1
XM_005259436.3 Intron XP_005259493.1
XM_011527542.2 Intron XP_011525844.1
XM_011527543.2 Intron XP_011525845.1
XM_011527544.2 Intron XP_011525846.1
XM_017027513.1 Intron XP_016883002.1
Gene
SLC6A16
Gene Name
solute carrier family 6 member 16
There are no transcripts associated with this gene.

Gene
TEAD2
Gene Name
TEA domain transcription factor 2
There are no transcripts associated with this gene.

View Full Product Details