Product Details

SNP ID
rs10883859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103368377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTGGAGGAGGCAGTGGCGGGC[G/T]CCGGAGCCCCGGGAGAGGTGGACAG
Phenotype
MIM: 601787
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TAF5 PubMed Links

Gene Details

Gene
TAF5
Gene Name
TATA-box binding protein associated factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006951.4 425 Missense Mutation GCC,TCC A130S NP_008882.2
NM_139052.2 425 Missense Mutation GCC,TCC A130S NP_620640.1

View Full Product Details