Product Details

SNP ID
rs11078354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16926835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTGGCCTGTGGGGCCCAGGGC[C/T]TATGGGGAGGAGCAGAGGGTGTCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBC1D27 PubMed Links

Gene Details

Gene
TBC1D27
Gene Name
TBC1 domain family member 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017025506.1 Intron XP_016880995.1

View Full Product Details