Product Details

SNP ID
rs11794020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:109136918 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATATCCAAAAGCACTTCTGCTTTA[C/T]GGAAATTTGAAATTTCTTCTGCCCA
Phenotype
MIM: 604574
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRRS1L PubMed Links

Gene Details

Gene
FRRS1L
Gene Name
ferric chelate reductase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011518453.2 1956 UTR 3 XP_011516755.1
XM_011518454.2 1956 UTR 3 XP_011516756.1

View Full Product Details