Product Details

SNP ID
rs10979727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:109174508 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACACAAAGCCCGAAGAAAGAAGA[C/T]GGACAGAAGGCACCATGCCATCTTC
Phenotype
MIM: 610340 MIM: 604574
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPB41L4B PubMed Links

Gene Details

Gene
EPB41L4B
Gene Name
erythrocyte membrane protein band 4.1 like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018424.3 2887 Intron NP_060894.2
NM_019114.4 2887 UTR 3 NP_061987.3
XM_011518790.1 2887 UTR 3 XP_011517092.1
XM_011518791.2 2887 UTR 3 XP_011517093.1
XM_011518792.2 2887 UTR 3 XP_011517094.1
XM_011518794.2 2887 UTR 3 XP_011517096.1
XM_017014813.1 2887 UTR 3 XP_016870302.1
XM_017014814.1 2887 UTR 3 XP_016870303.1
XM_017014815.1 2887 UTR 3 XP_016870304.1
Gene
FRRS1L
Gene Name
ferric chelate reductase 1 like
There are no transcripts associated with this gene.

View Full Product Details