Product Details

SNP ID
rs10816774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:109174721 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCCCAGGTTCTTTCCTGATCTCA[A/G]TGTTGGACTTTCTTTTTTAGACTCC
Phenotype
MIM: 610340 MIM: 604574
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPB41L4B PubMed Links

Gene Details

Gene
EPB41L4B
Gene Name
erythrocyte membrane protein band 4.1 like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018424.3 Intron NP_060894.2
NM_019114.4 Intron NP_061987.3
XM_011518790.1 Intron XP_011517092.1
XM_011518791.2 Intron XP_011517093.1
XM_011518792.2 Intron XP_011517094.1
XM_011518794.2 Intron XP_011517096.1
XM_017014813.1 Intron XP_016870302.1
XM_017014814.1 Intron XP_016870303.1
XM_017014815.1 Intron XP_016870304.1
Gene
FRRS1L
Gene Name
ferric chelate reductase 1 like
There are no transcripts associated with this gene.

View Full Product Details