Product Details
- SNP ID
-
rs3766359
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
47
- Location
-
Chr.1:160879270 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- CATACCAACACACACGGACAAACTC[A/G]ACCTTACTCACAGGTCCCTGCAGTC
- Phenotype
-
MIM: 609873
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ITLN1
PubMed Links
Gene Details
- Gene
- ITLN1
- Gene Name
- intelectin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017625.2 |
|
Intron |
|
|
NP_060095.2 |
View Full Product Details