Product Details
- SNP ID
-
rs3766356
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
46
- Location
-
Chr.1:160879147 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- ATGTATACCTGGACAAAAGTTCCCA[C/T]GGCCCTGGCCCCTGAAGCTCAGCAA
- Phenotype
-
MIM: 609873
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ITLN1
PubMed Links
Gene Details
- Gene
- ITLN1
- Gene Name
- intelectin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017625.2 |
|
Intron |
|
|
NP_060095.2 |
View Full Product Details