Product Details
- SNP ID
-
rs11841945
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:73055921 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACTCTATACCTCTTAATTTCAAGCT[C/G]TGACGTTTTTGCCAAACAACTGAAT
- Phenotype
-
MIM: 602903
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KLF5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61966249] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLF5
- Gene Name
- Kruppel like factor 5
View Full Product Details