Product Details

SNP ID
rs11206831
Assay Type
Validated
NCBI dbSNP Submissions
30
Location
Chr.1:56497354 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACTTGAGAAACATTTATTGAGTGC[C/T]ACTTGCAAAGGTGTGAGGATGGCAC
Phenotype
MIM: 607125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLPP3 PubMed Links

Gene Details

Gene
PLPP3
Gene Name
phospholipid phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003713.4 Intron NP_003704.3

View Full Product Details