Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001010881.1 | 6685 | Intron | NP_001010881.1 | ||
XM_011541269.2 | 6685 | Intron | XP_011539571.2 | ||
XM_011541271.2 | 6685 | Intron | XP_011539573.2 | ||
XM_011541272.2 | 6685 | Intron | XP_011539574.1 | ||
XM_011541274.2 | 6685 | Intron | XP_011539576.2 | ||
XM_011541275.2 | 6685 | Intron | XP_011539577.2 | ||
XM_011541276.2 | 6685 | Intron | XP_011539578.1 | ||
XM_011541277.2 | 6685 | Intron | XP_011539579.1 | ||
XM_011541278.2 | 6685 | Intron | XP_011539580.1 | ||
XM_011541280.2 | 6685 | Intron | XP_011539582.2 | ||
XM_011541281.2 | 6685 | Intron | XP_011539583.2 | ||
XM_017001062.1 | 6685 | Intron | XP_016856551.1 | ||
XM_017001063.1 | 6685 | Intron | XP_016856552.1 | ||
XM_017001064.1 | 6685 | Intron | XP_016856553.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005957.4 | 6685 | UTR 3 | NP_005948.3 | ||
XM_005263458.3 | 6685 | Intron | XP_005263515.1 | ||
XM_005263460.4 | 6685 | Intron | XP_005263517.1 | ||
XM_005263462.4 | 6685 | Intron | XP_005263519.1 | ||
XM_005263463.3 | 6685 | Intron | XP_005263520.1 | ||
XM_011541495.2 | 6685 | Intron | XP_011539797.1 | ||
XM_011541496.2 | 6685 | Intron | XP_011539798.1 | ||
XM_017001328.1 | 6685 | Intron | XP_016856817.1 |