Product Details

SNP ID
rs7329109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:71445720 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATTAAAACATACTTGCAGATCA[A/T]TAAAAGTCTGTAGGAACAAAATTTT
Phenotype
MIM: 603803
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DACH1 PubMed Links

Gene Details

Gene
DACH1
Gene Name
dachshund family transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004392.6 Intron NP_004383.4
NM_080759.5 Intron NP_542937.3
NM_080760.5 Intron NP_542938.3
XM_011534939.2 Intron XP_011533241.1
XM_011534940.2 Intron XP_011533242.1
XM_011534941.2 Intron XP_011533243.1
XM_011534942.2 Intron XP_011533244.1
XM_017020396.1 Intron XP_016875885.1

View Full Product Details