Product Details

SNP ID
rs6573875
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69193040 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACTGCCACCCACGGTGGATAATA[A/C]TTTCCTCCATTCTCTCTCTCTTTTT
Phenotype
MIM: 616940
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
EXD2 PubMed Links

Gene Details

Gene
EXD2
Gene Name
exonuclease 3'-5' domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193360.1 Intron NP_001180289.1
NM_001193361.1 Intron NP_001180290.1
NM_001193362.1 Intron NP_001180291.1
NM_001193363.1 Intron NP_001180292.1
NM_018199.3 Intron NP_060669.1
XM_005267817.3 Intron XP_005267874.1
XM_011536908.2 Intron XP_011535210.1
XM_017021421.1 Intron XP_016876910.1
Gene
LOC105370550
Gene Name
uncharacterized LOC105370550
There are no transcripts associated with this gene.

View Full Product Details