Product Details
- SNP ID
-
rs3812694
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:70255817 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGCTTCTCGCGGAAAGGGTGCACGA[G/T]GCAGCGGAACCTGCCGCGGGGAGAG
- Phenotype
-
MIM: 607448
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NPFFR1
PubMed Links
Gene Details
- Gene
- NPFFR1
- Gene Name
- neuropeptide FF receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022146.4 |
451 |
Missense Mutation |
ATC,CTC |
I145L |
NP_071429.1 |
View Full Product Details