Product Details

SNP ID
rs10999212
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:70255653 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCCAGGCCTCCCAGCAGGAGTAGAG[A/C]GGGTAGGAGCGGTTGCGGGCGTCCA
Phenotype
MIM: 607448
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NPFFR1 PubMed Links

Gene Details

Gene
NPFFR1
Gene Name
neuropeptide FF receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022146.4 615 Silent Mutation NP_071429.1

View Full Product Details