Product Details

SNP ID
rs1717774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:30328638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTGTATTTTATTTTTTAACTTT[C/G]TAAATACAGTATTTTGACCTAAAAC
Phenotype
MIM: 612295
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL14EP PubMed Links

Gene Details

Gene
ARL14EP
Gene Name
ADP ribosylation factor like GTPase 14 effector protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152316.2 Intron NP_689529.1
XM_005252792.3 Intron XP_005252849.1

View Full Product Details