Product Details

SNP ID
rs9367279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:47480600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAGTTTCTCAATCTATAAATGGA[A/G]TAAAATATCAACCTCATAGGTTGTT
Phenotype
MIM: 604241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD2AP PubMed Links

Gene Details

Gene
CD2AP
Gene Name
CD2 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012120.2 Intron NP_036252.1
XM_005248976.1 Intron XP_005249033.1
XM_011514449.2 Intron XP_011512751.1
XM_017010641.1 Intron XP_016866130.1

View Full Product Details