Product Details

SNP ID
rs10858185
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:135937699 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCGCGGCCACGGCGACGGCCACACC[A/G]GAGGGAGGGATGAGGTGCATAGGGG
Phenotype
MIM: 608129
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBAC1 PubMed Links

Gene Details

Gene
UBAC1
Gene Name
UBA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016172.2 Intron NP_057256.2

View Full Product Details