Product Details

SNP ID
rs35099862
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:123982657 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CACCGTATCGGTTTTACTGTTGCAG[A/C]GCTTACAGGAGCAGATTCATTTAGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC92 PubMed Links

Gene Details

Gene
CCDC92
Gene Name
coiled-coil domain containing 92
There are no transcripts associated with this gene.

Gene
ZNF664
Gene Name
zinc finger protein 664
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204298.1 Intron NP_001191227.1
NM_152437.2 Intron NP_689650.1
Gene
ZNF664-FAM101A
Gene Name
filamin-interacting protein FAM101A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204299.1 Intron NP_001191228.1

View Full Product Details