Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_032840.2 | 2641 | UTR 3 | NP_116229.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015319.2 | 2641 | Intron | NP_056134.2 | ||
NM_170754.2 | 2641 | Intron | NP_736610.2 | ||
NM_198316.1 | 2641 | Intron | NP_938072.1 | ||
XM_006719302.3 | 2641 | Intron | XP_006719365.1 | ||
XM_006719303.1 | 2641 | Intron | XP_006719366.1 | ||
XM_011538079.1 | 2641 | Intron | XP_011536381.1 | ||
XM_017019088.1 | 2641 | Intron | XP_016874577.1 | ||
XM_017019089.1 | 2641 | Intron | XP_016874578.1 |