Product Details

SNP ID
rs35551560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196218314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTGGAGGGCTGCACGGCATCC[A/G]CCCTGGACCCATGTCACTCTAGAGG
Phenotype
MIM: 612084
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC51A PubMed Links

Gene Details

Gene
SLC51A
Gene Name
solute carrier family 51 alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152672.5 Intron NP_689885.4
Gene
ZDHHC19
Gene Name
zinc finger DHHC-type containing 19
There are no transcripts associated with this gene.

View Full Product Details