Product Details

SNP ID
rs2341097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45765644 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTCGGGGTCTGGCAGCCTCAGCA[C/T]GGTGTGGGGGGCCTGTGTCCCCAGC
Phenotype
MIM: 605377 MIM: 600963
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BHMG1 PubMed Links

Gene Details

Gene
BHMG1
Gene Name
basic helix-loop-helix and HMG-box containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310124.1 2472 Intron NP_001297053.1
XM_011527571.2 2472 Intron XP_011525873.1
XM_011527573.2 2472 Intron XP_011525875.1
XM_011527574.2 2472 Intron XP_011525876.1
Gene
DMPK
Gene Name
dystrophia myotonica protein kinase
There are no transcripts associated with this gene.

Gene
SIX5
Gene Name
SIX homeobox 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175875.4 2472 Missense Mutation ATG,GTG M693V NP_787071.2

View Full Product Details