Product Details

SNP ID
rs2301287
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:161167053 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CTTCCCCTCCCCTCCTGACCTCTCG[C/G]CGGCGGCTACAGGCGGTGCTGCAGT
Phenotype
MIM: 604014 MIM: 600923 MIM: 610554 MIM: 604729
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
B4GALT3 PubMed Links

Gene Details

Gene
B4GALT3
Gene Name
beta-1,4-galactosyltransferase 3
There are no transcripts associated with this gene.

Gene
PPOX
Gene Name
protoporphyrinogen oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000309.3 Intron NP_000300.1
NM_001122764.1 Intron NP_001116236.1
XM_005245291.3 Intron XP_005245348.2
XM_005245295.3 Intron XP_005245352.2
XM_006711402.2 Intron XP_006711465.2
XM_006711403.2 Intron XP_006711466.2
XM_006711404.3 Intron XP_006711467.2
XM_006711406.2 Intron XP_006711469.2
XM_011509663.2 Intron XP_011507965.1
XM_011509664.1 Intron XP_011507966.1
XM_011509665.2 Intron XP_011507967.1
XM_011509666.2 Intron XP_011507968.1
XM_011509667.2 Intron XP_011507969.1
XM_011509668.2 Intron XP_011507970.1
XM_011509669.1 Intron XP_011507971.1
XM_011509670.2 Intron XP_011507972.1
XM_011509671.1 Intron XP_011507973.1
XM_011509672.2 Intron XP_011507974.1
XM_011509673.2 Intron XP_011507975.1
XM_011509674.2 Intron XP_011507976.1
XM_011509675.2 Intron XP_011507977.1
XM_011509676.1 Intron XP_011507978.1
XM_011509677.1 Intron XP_011507979.1
XM_011509678.1 Intron XP_011507980.1
XM_011509679.1 Intron XP_011507981.1
XM_011509681.1 Intron XP_011507983.1
XM_011509682.1 Intron XP_011507984.1
XM_017001559.1 Intron XP_016857048.1
XM_017001560.1 Intron XP_016857049.1
XM_017001561.1 Intron XP_016857050.1
XM_017001562.1 Intron XP_016857051.1
XM_017001563.1 Intron XP_016857052.1
XM_017001564.1 Intron XP_016857053.1
XM_017001565.1 Intron XP_016857054.1
XM_017001566.1 Intron XP_016857055.1
XM_017001567.1 Intron XP_016857056.1
XM_017001568.1 Intron XP_016857057.1
XM_017001569.1 Intron XP_016857058.1
XM_017001570.1 Intron XP_016857059.1
XM_017001571.1 Intron XP_016857060.1
Gene
UFC1
Gene Name
ubiquitin-fold modifier conjugating enzyme 1
There are no transcripts associated with this gene.

Gene
USP21
Gene Name
ubiquitin specific peptidase 21
There are no transcripts associated with this gene.

View Full Product Details