Product Details
- SNP ID
-
rs2344206
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:3470595 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTGGGGCCGGCTGGCTTGTGGAAG[A/G]TGTGAGAAGCCGTCGTGGCAAATTC
- Phenotype
-
MIM: 610285
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DOK7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74705472] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DOK7
- Gene Name
- docking protein 7
View Full Product Details