Product Details

SNP ID
rs11707899
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:186666226 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACTTGGACAGAGTGGTGAGGAATTG[C/T]CAATGGCAGAGCTGAGTTGGGAGAT
Phenotype
MIM: 142640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HRG PubMed Links

Gene Details

Gene
HRG
Gene Name
histidine rich glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000412.4 Intron NP_000403.1
XM_005247415.3 Intron XP_005247472.1

View Full Product Details