Product Details

SNP ID
rs7220169
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:41715448 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTTTGCAGACGAGATGCAGCGACT[A/G]TGTGTGTATGTGCTGATCTTTGCAC
Phenotype
MIM: 137250 MIM: 600947
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAST PubMed Links

Gene Details

Gene
GAST
Gene Name
gastrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000805.4 79 Silent Mutation CTA,CTG L4L NP_000796.1
Gene
HAP1
Gene Name
huntingtin associated protein 1
There are no transcripts associated with this gene.

View Full Product Details