Product Details
- SNP ID
-
rs9049
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:138494450 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTTAAATGAGAGACTTCTATCTA[C/T]TCATCCATTTTACCCTATGATTCAT
- Phenotype
-
MIM: 614310
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP70
PubMed Links
Gene Details
- Gene
- CEP70
- Gene Name
- centrosomal protein 70
View Full Product Details