Product Details

SNP ID
rs35656218
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:1056856 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTGTGGGGTCGCGTCAGTGCGGAGA[C/G]AGCGGTGTCCTGCGCGTCTTCGGGT
Phenotype
MIM: 604055
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IDI1 PubMed Links

Gene Details

Gene
IDI1
Gene Name
isopentenyl-diphosphate delta isomerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317955.1 21 Intron NP_001304884.1
NM_001317956.1 21 Intron NP_001304885.1
NM_001317957.1 21 Intron NP_001304886.1
NM_004508.3 21 Intron NP_004499.2
XM_011519457.1 21 Intron XP_011517759.1
XM_017016191.1 21 Intron XP_016871680.1
Gene
WDR37
Gene Name
WD repeat domain 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014023.3 21 UTR 5 NP_054742.2

View Full Product Details