Product Details

SNP ID
rs1046053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59659798 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAAATGATTTTCATTTGGTGCTTA[A/C]GTGCTTTATGTGAATTTTTCATAGC
Phenotype
MIM: 603292 MIM: 600519
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BNIP2 PubMed Links

Gene Details

Gene
BNIP2
Gene Name
BCL2 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320674.1 5709 UTR 3 NP_001307603.1
NM_001320675.1 5709 UTR 3 NP_001307604.1
NM_004330.3 5709 UTR 3 NP_004321.2
Gene
GTF2A2
Gene Name
general transcription factor IIA subunit 2
There are no transcripts associated with this gene.

View Full Product Details