Product Details

SNP ID
rs4795048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34928743 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCTTTCCCCCTCAAATAGTAAG[A/G]TTAATTTTTCTTAGAACTTAACACT
Phenotype
MIM: 610730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCT6B PubMed Links

Gene Details

Gene
CCT6B
Gene Name
chaperonin containing TCP1 subunit 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193529.2 Intron NP_001180458.1
NM_001193530.1 Intron NP_001180459.1
NM_006584.3 Intron NP_006575.2
XM_011524203.2 Intron XP_011522505.1
XM_017024023.1 Intron XP_016879512.1
XM_017024024.1 Intron XP_016879513.1

View Full Product Details