Product Details

SNP ID
rs11993938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:53729613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCACCCTGCTGTGGGGAGGAAGG[C/G]AGTGTGGAATGGAGCCCAGGCTATT
Phenotype
MIM: 608861
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP6V1H PubMed Links

Gene Details

Gene
ATP6V1H
Gene Name
ATPase H+ transporting V1 subunit H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015941.3 Intron NP_057025.2
NM_213619.2 Intron NP_998784.1
NM_213620.2 Intron NP_998785.1
XM_006716455.2 Intron XP_006716518.1
XM_011517542.1 Intron XP_011515844.1

View Full Product Details