Product Details

SNP ID
rs2761210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33385425 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCATTAGAAGTTTAGCAATATTT[A/C]TGACTGCCTGGTAGATGTCAGTAAC
Phenotype
MIM: 612297
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1549L PubMed Links

Gene Details

Gene
KIAA1549L
Gene Name
KIAA1549 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012194.2 Intron NP_036326.2
XM_005252847.3 Intron XP_005252904.2
XM_005252848.3 Intron XP_005252905.1
XM_011519969.2 Intron XP_011518271.1
XM_011519970.2 Intron XP_011518272.1
XM_017017484.1 Intron XP_016872973.1
XM_017017485.1 Intron XP_016872974.1
XM_017017486.1 Intron XP_016872975.1

View Full Product Details