Product Details

SNP ID
rs12681420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:70680963 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAGGGAGTAGAAATTAAAATTCA[A/G]TGCTATACCAAAGGGTATACTAATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XKR9 PubMed Links

Gene Details

Gene
XKR9
Gene Name
XK related 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011720.1 422 UTR 5 NP_001011720.1
NM_001287258.1 422 UTR 5 NP_001274187.1
NM_001287259.1 422 UTR 5 NP_001274188.1
NM_001287260.1 422 UTR 5 NP_001274189.1
XM_006716447.3 422 Intron XP_006716510.1
XM_011517524.2 422 UTR 5 XP_011515826.1
XM_011517525.2 422 UTR 5 XP_011515827.1
XM_011517526.2 422 UTR 5 XP_011515828.1
XM_011517527.2 422 UTR 5 XP_011515829.1
XM_017013404.1 422 UTR 5 XP_016868893.1
XM_017013405.1 422 UTR 5 XP_016868894.1

View Full Product Details