Product Details

SNP ID
rs2296135
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:5952731 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTATGTCACTTTTCTCTGTGAACTG[A/C]AAGTTAGGATGAGGGACGGAAGATT
Phenotype
MIM: 601070
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL15RA PubMed Links

Gene Details

Gene
IL15RA
Gene Name
interleukin 15 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243539.1 1681 UTR 3 NP_001230468.1
NM_001256765.1 1681 UTR 3 NP_001243694.1
NM_002189.3 1681 UTR 3 NP_002180.1
NM_172200.2 1681 UTR 3 NP_751950.2
XM_011519461.2 1681 Intron XP_011517763.1
XM_011519462.2 1681 Intron XP_011517764.1
XM_011519463.2 1681 Intron XP_011517765.1
XM_011519464.2 1681 Intron XP_011517766.1
XM_011519465.1 1681 UTR 3 XP_011517767.1
XM_011519466.2 1681 UTR 3 XP_011517768.1
XM_011519467.2 1681 Intron XP_011517769.1
XM_011519468.1 1681 Intron XP_011517770.1
XM_011519469.1 1681 UTR 3 XP_011517771.1
XM_011519470.2 1681 Intron XP_011517772.1
XM_011519471.2 1681 Intron XP_011517773.1
XM_011519472.2 1681 Intron XP_011517774.1
XM_011519474.2 1681 Intron XP_011517776.1
XM_011519475.2 1681 Intron XP_011517777.1
XM_011519476.2 1681 UTR 3 XP_011517778.1
XM_011519477.2 1681 UTR 3 XP_011517779.1
XM_017016195.1 1681 UTR 3 XP_016871684.1
XM_017016196.1 1681 Intron XP_016871685.1
XM_017016197.1 1681 Intron XP_016871686.1
XM_017016198.1 1681 UTR 3 XP_016871687.1
XM_017016199.1 1681 Intron XP_016871688.1
XM_017016200.1 1681 Intron XP_016871689.1
XM_017016201.1 1681 UTR 3 XP_016871690.1
XM_017016202.1 1681 UTR 3 XP_016871691.1
XM_017016203.1 1681 UTR 3 XP_016871692.1

View Full Product Details