Product Details

SNP ID
rs1532834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55022144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAGCAAAGACAATCTTATTTTC[A/G]TTTTGCCCTAAGGGTACAAAAATTA
Phenotype
MIM: 611635
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEUROD4 PubMed Links

Gene Details

Gene
NEUROD4
Gene Name
neuronal differentiation 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021191.2 Intron NP_067014.2

View Full Product Details