Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004842.3 | Intron | NP_004833.1 | |||
NM_016377.3 | Intron | NP_057461.2 | |||
NM_138633.2 | Intron | NP_619539.1 | |||
XM_005267228.1 | Intron | XP_005267285.1 | |||
XM_005267229.4 | Intron | XP_005267286.1 | |||
XM_006715614.2 | Intron | XP_006715677.1 | |||
XM_011536262.1 | Intron | XP_011534564.1 | |||
XM_017011506.1 | Intron | XP_016866995.1 | |||
XM_017011507.1 | Intron | XP_016866996.1 | |||
XM_017011508.1 | Intron | XP_016866997.1 | |||
XM_017011509.1 | Intron | XP_016866998.1 | |||
XM_017011510.1 | Intron | XP_016866999.1 | |||
XM_017011511.1 | Intron | XP_016867000.1 |