Product Details

SNP ID
rs11802159
Assay Type
Functionally Tested
NCBI dbSNP Submissions
16
Location
Chr.1:183647957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGTTGGGTTGGAGTTGTCCACT[C/T]GGCATTTGAAAAAACTGTCCAGGAA
Phenotype
MIM: 609908 MIM: 605667
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
APOBEC4 PubMed Links
Additional Information
For this assay, SNP(s) [rs10911391] are located under a probe and SNP(s) [rs1174658] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
APOBEC4
Gene Name
apolipoprotein B mRNA editing enzyme catalytic polypeptide like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203454.2 1097 Silent Mutation CCA,CCG P275P NP_982279.1
Gene
RGL1
Gene Name
ral guanine nucleotide dissociation stimulator like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297669.1 1097 Intron NP_001284598.1
NM_001297670.1 1097 Intron NP_001284599.1
NM_001297671.1 1097 Intron NP_001284600.1
NM_001297672.1 1097 Intron NP_001284601.1
NM_015149.4 1097 Intron NP_055964.3
XM_011509339.2 1097 Intron XP_011507641.1
XM_011509341.1 1097 Intron XP_011507643.1
XM_011509342.2 1097 Intron XP_011507644.1
XM_017000756.1 1097 Intron XP_016856245.1

View Full Product Details