Product Details

SNP ID
rs402691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53888383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCAGATATTGTCCCGGGGGTAG[C/T]GGGTGGCAGGGGAGAAGGAGAAGCT
Phenotype
MIM: 176980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKCG PubMed Links

Gene Details

Gene
PRKCG
Gene Name
protein kinase C gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316329.1 Intron NP_001303258.1
NM_002739.4 Intron NP_002730.1

View Full Product Details