Product Details

SNP ID
rs4901637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:56120242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGTAACTTCAGCCCAACCTAAT[C/T]TTGTGGAGTTTGGTTTTGTTTGTGT
Phenotype
MIM: 614798
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PELI2 PubMed Links

Gene Details

Gene
PELI2
Gene Name
pellino E3 ubiquitin protein ligase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021255.2 Intron NP_067078.1
XM_005267890.4 Intron XP_005267947.1
XM_006720211.3 Intron XP_006720274.1
XM_011536990.2 Intron XP_011535292.1
XM_011536992.2 Intron XP_011535294.1
XM_017021478.1 Intron XP_016876967.1
XM_017021479.1 Intron XP_016876968.1

View Full Product Details