Product Details

SNP ID
rs813943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104032447 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTTAACTTGTTTGCCCCATTTC[A/C]ACACCAGCTTGGTTCTCCTTTTCCT
Phenotype
MIM: 113811 MIM: 616563
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COL17A1 PubMed Links

Gene Details

Gene
COL17A1
Gene Name
collagen type XVII alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000494.3 Intron NP_000485.3
Gene
SLK
Gene Name
STE20 like kinase
There are no transcripts associated with this gene.

View Full Product Details