Product Details

SNP ID
rs3782126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57463150 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCCAAGAAGATCCCCAGAGTACA[C/T]AGACTACAAGACTGCCTCTGCCTCT
Phenotype
MIM: 610576 MIM: 165220 MIM: 612031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP9 PubMed Links

Gene Details

Gene
ARHGAP9
Gene Name
Rho GTPase activating protein 9
There are no transcripts associated with this gene.

Gene
GLI1
Gene Name
GLI family zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160045.1 Intron NP_001153517.1
NM_001167609.1 Intron NP_001161081.1
NM_005269.2 Intron NP_005260.1
XM_011538189.2 Intron XP_011536491.1
XM_011538190.1 Intron XP_011536492.1
Gene
INHBE
Gene Name
inhibin beta E subunit
There are no transcripts associated with this gene.

View Full Product Details