Product Details

SNP ID
rs9525563
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:42046848 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTTGACTTTGGTCTCAGTTTGTCA[A/G]TTGGGGCCTTAGTTTCTTTACATTA
Phenotype
MIM: 604071
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKH PubMed Links

Gene Details

Gene
DGKH
Gene Name
diacylglycerol kinase eta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204504.2 Intron NP_001191433.1
NM_001204505.2 Intron NP_001191434.1
NM_001204506.2 Intron NP_001191435.1
NM_001297429.1 Intron NP_001284358.1
NM_152910.5 Intron NP_690874.2
NM_178009.4 Intron NP_821077.1

View Full Product Details