Product Details

SNP ID
rs4759276
Assay Type
Functionally Tested
NCBI dbSNP Submissions
38
Location
Chr.12:57132863 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGGGCTTTCCCACTCACCCCCT[A/G]GCTCCCTCCCCTCTGTTTTGGCTAT
Phenotype
MIM: 107770
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LRP1 PubMed Links

Gene Details

Gene
LRP1
Gene Name
LDL receptor related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002332.2 Intron NP_002323.2
XM_017019303.1 Intron XP_016874792.1

View Full Product Details