Product Details

SNP ID
rs13179664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:126549242 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTGAACTGAAAACAGCAAAGTCC[A/G]TGCATCAAATAGTAATTGTAGTATC
Phenotype
MIM: 107323
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH7A1 PubMed Links

Gene Details

Gene
ALDH7A1
Gene Name
aldehyde dehydrogenase 7 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001182.4 Intron NP_001173.2
NM_001201377.1 Intron NP_001188306.1
NM_001202404.1 Intron NP_001189333.2
XM_011543417.2 Intron XP_011541719.1
XM_017009493.1 Intron XP_016864982.1

View Full Product Details