Product Details

SNP ID
rs1454164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:176324805 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGAAGATAATGCTAATAATCCAT[A/G]CATATGTAAACAGTAGGAAAATGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPCS3 PubMed Links

Gene Details

Gene
SPCS3
Gene Name
signal peptidase complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021928.3 Intron NP_068747.1

View Full Product Details