Product Details

SNP ID
rs10893435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125838890 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAAGATGCAAGTAAAGGGAGATA[C/T]GACAAAGAAGAGGAAAAGATGATGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PATE4 PubMed Links

Gene Details

Gene
PATE4
Gene Name
prostate and testis expressed 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144874.1 804 UTR 3 NP_001138346.1

View Full Product Details