Product Details

SNP ID
rs923118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74173930 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCCAAGTCCTCCCTTGGGCGCCC[G/T]TGGCCCTGCAGACTCTCAGGGCTAA
Phenotype
MIM: 602059 MIM: 610745
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ISLR PubMed Links

Gene Details

Gene
ISLR
Gene Name
immunoglobulin superfamily containing leucine rich repeat
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005545.3 185 UTR 5 NP_005536.1
NM_201526.1 185 Intron NP_958934.1
Gene
STRA6
Gene Name
stimulated by retinoic acid 6
There are no transcripts associated with this gene.

View Full Product Details