Product Details

SNP ID
rs12011018
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:2849258 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTACGCAACTGTGGTGAAGATGCC[A/G]TAAATTCCATAAATAACTGGGCCAA
Phenotype
MIM: 300198
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GYG2 PubMed Links

Gene Details

Gene
GYG2
Gene Name
glycogenin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079855.1 Intron NP_001073324.1
NM_001184702.1 Intron NP_001171631.1
NM_001184703.1 Intron NP_001171632.1
NM_001184704.1 Intron NP_001171633.1
NM_003918.2 Intron NP_003909.2
XM_011545600.2 Intron XP_011543902.1
XM_017029927.1 Intron XP_016885416.1
XM_017029928.1 Intron XP_016885417.1
XM_017029929.1 Intron XP_016885418.1
XM_017029930.1 Intron XP_016885419.1
XM_017029931.1 Intron XP_016885420.1

View Full Product Details